missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIAA0319L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567257
This item is not returnable.
View return policy
Description
Immunogen sequence: KRKSKYKILDA TDQESLELKP TSRAGIKQKG LLLSSSLMHS ESELDSDDAI FTWPDREKGK LLHGQNGSVP NGQ Highest antigen sequence identity to the following orthologs - mouse 93%, rat 93%.
Possible role in axon guidance through interaction with RTN4R. (Microbial infection) Acts as a receptor for adeno-associated virus and is involved in adeno-associated virus infection through endocytosis system.
Specifications
| KIAA0319L | |
| Polyclonal | |
| Unconjugated | |
| KIAA0319L | |
| A730047D20Rik; AAV receptor; AAVR; Adeno-associated virus receptor; AU040320; Dyslexia-associated protein KIAA0319-like protein; dyslexia-associated protein KIAA0319-like protein homolog; expressed sequence AU040320; KIAA0319 like; Kiaa0319l; KIAA0319-like; KIAA0319-like ortholog; KIAA1837; likely ortholog of H. sapiens polycystic kidney disease 1-like (PKD1-like); LOW QUALITY PROTEIN: dyslexia-associated protein KIAA0319-like protein homolog; polycystic kidney disease 1-related; PP791 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79932 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IZA0 | |
| KIAA0319L | |
| Recombinant Human KIAA0319L. Recombinant protein control fragment (Product #RP-107893). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction