missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIAA0317 Monoclonal Antibody (K56023_6E4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555638
This item is not returnable.
View return policy
Description
Sequence of this protein is as follows: FDIIVLSEDE KNIVERNVST SGVSIYFEAY LYNATNCSST PWHLPPMHMT SSQRRPSTAV DEEDEDSPSE CHTPEKVKKP
KIAA0317 contains 1 filamin repeat and 1 HECT (E6AP-type E3 ubiquitin-protein ligase) domain. The exact function of KIAA0317 remains unknown.
Specifications
| KIAA0317 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| O15033 | |
| AREL1 | |
| Recombinant human AREL1 (immunogen range is 271-350). | |
| 50 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2a |
| Immunoprecipitation, Western Blot | |
| K56023_6E4 | |
| Unconjugated | |
| AREL1 | |
| apoptosis resistant E3 ubiquitin protein ligase 1; apoptosis-resistant E3 ubiquitin protein ligase 1; Apoptosis-resistant HECT-type E3 ubiquitin transferase 1; AREL1; fibrosis-inducing E3 ligase 1; FIEL1; KIAA0317; protein KIAA0317 | |
| Mouse | |
| Protein A | |
| RUO | |
| 299197, 9870 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction