missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA0284 Mouse anti-Human, Clone: 1D4B8, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | KIAA0284 |
|---|---|
| Clone | 1D4B8 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16828915
|
Proteintech
66749-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16818915
|
Proteintech
66749-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CEP170B plays a role in microtubule organization.Specifications
| KIAA0284 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Cep170B, FAM68C, KIAA0284, Protein KIAA0284 | |
| CEP170B | |
| IgG2b | |
| Protein A | |
| Antibody |
| 1D4B8 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9Y4F5 | |
| 283638 | |
| KIAA0284 Fusion Protein Ag26852 | |
| Primary | |
| -20°C | |
| CEP170B |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title