missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIAA0100 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549826
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total KIAA0100 / BCOX1 protein.
Belongs to the UPF0378 family. 4 isoforms of the human protein are produced by alternative splicing. Song J., Biochim. Biophys. Acta 1760:62-69(2006). Nagase T., DNA Res. 2:37-43(1995). Chen G., Leuk. Res. 29:503-509(2005).
Specifications
| KIAA0100 | |
| Polyclonal | |
| Unconjugated | |
| KIAA0100 | |
| Antigen MLAA-22; BCOX; BCOX1; breast cancer overexpressed gene 1; breast cancer-overexpressed gene 1 protein; cancer/testis antigen 101; CT101; FMP27; K0100; KIAA0100; protein KIAA0100; U937-associated antigen | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9703 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14667 | |
| KIAA0100 | |
| Synthesized peptide derived from internal of human K0100. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction