missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA0090 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26017-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a single-pass type I transmembrane protein, which is a subunit of the endoplasmic reticulum membrane protein complex (EMC). Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| KIAA0090 | |
| Polyclonal | |
| Unconjugated | |
| EMC1 | |
| EMC1, ER membrane protein complex subunit 1, KIAA0090 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23065 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8N766 | |
| EMC1 | |
| KIAA0090 Fusion Protein Ag23406 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction