missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIAA0020 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531247
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (87%), Rat (86%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Inhibits the poly(ADP-ribosyl)ation activity of PARP1 and the degradation of PARP1 by CASP3 following genotoxic stress. Binds to double-stranded RNA or DNA without sequence specificity. Involved in development of the eye and of primordial germ cells.
Specifications
| KIAA0020 | |
| Polyclonal | |
| Unconjugated | |
| PUM3 | |
| cPERP-C; HA-8; HBV XAg-transactivated protein 5; HBV X-transactivated gene 5 protein; HLA-HA8; KIAA0020; Minor histocompatibility antigen HA-8; PEN; penguin homolog; protein 5 transactivated by hepatitis B virus X antigen (HBxAg); PUF6; PUF-A; PUM3; Pumilio homolog 3; pumilio RNA binding family member 3; pumilio RNA-binding family member 3; XTP5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9933 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q15397 | |
| PUM3 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human KIAA0020. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction