missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KIAA0020 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527851
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Recommended positive controls: 293T, A431, Jurkat, Raji. Predicted reactivity: Mouse (88%), Rat (87%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Inhibits the poly(ADP-ribosyl)ation activity of PARP1 and the degradation of PARP1 by CASP3 following genotoxic stress. Binds to double-stranded RNA or DNA without sequence specificity. Involved in development of the eye and of primordial germ cells.
Spécification
| KIAA0020 | |
| Polyclonal | |
| Unconjugated | |
| PUM3 | |
| cPERP-C; HA-8; HBV XAg-transactivated protein 5; HBV X-transactivated gene 5 protein; HLA-HA8; KIAA0020; Minor histocompatibility antigen HA-8; PEN; penguin homolog; protein 5 transactivated by hepatitis B virus X antigen (HBxAg); PUF6; PUF-A; PUM3; Pumilio homolog 3; pumilio RNA binding family member 3; pumilio RNA-binding family member 3; XTP5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9933 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q15397 | |
| PUM3 | |
| Recombinant protein encompassing a sequence within the center region of human KIAA0020. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu