missing translation for 'onlineSavingsMsg'
Learn More

KI696, MedChemExpress

Product Code. 16474536
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16474536 100 mg 100mg
16460729 1 mg 1mg
16436007 5 mg 5mg
16432706 25 mg 25mg
16429826 50 mg 50mg
16413288 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16474536 Supplier Medchem Express Supplier No. HY101140

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress KI696 is a high affinity probe that disrupts the Keap1/NRF2 interaction. KI696 is a potent and selective inhibitor of the KEAP1/NRF2 interaction.

KI696 is a high affinity probe that disrupts the Keap1/NRF2 interaction. KI696 is a potent and selective inhibitor of the KEAP1/NRF2 interaction.

Specifications

Chemical Name or Material KI696
CAS 1799974-70-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C28H30N4O6S
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (90.81 mM; Need ultrasonic)
SMILES COC1=C(N2C)C(N=N2)=CC([C@H](C3=CC(CN(C[C@H]4C)S(=O)(C5=CC=CC=C5O4)=O)=C(C)C=C3)CC(O)=O)=C1
Molecular Weight (g/mol) 550.63
Formula Weight 550.63
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.