missing translation for 'onlineSavingsMsg'
Learn More

KI696 isomer, MedChemExpress

Product Code. 16470729
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16470729 1 mg 1mg
16446007 5 mg 5mg
16442706 25 mg 25mg
16423288 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16470729 Supplier Medchem Express Supplier No. HY101140A
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress KI696 isomer is the less active isomer of KI696 (HY-101140). KI696 is a high affinity probe that disrupts the Keap1/NRF2 interaction.

KI696 isomer is the less active isomer of KI696 (HY-101140). KI696 is a high affinity probe that disrupts the Keap1/NRF2 interaction.

Specifications

Chemical Name or Material KI696 isomer
CAS 1799974-69-8
Color White
Purity Grade Notes Research
Molecular Formula C28H30N4O6S
Quantity 1 mg
Solubility Information DMSO : 125 mg/mL (227.01 mM; Need ultrasonic)
SMILES COC1=C(N2C)C(N=N2)=CC([C@@H](C3=CC(CN(C[C@H]4C)S(=O)(C5=CC=CC=C5O4)=O)=C(C)C=C3)CC(O)=O)=C1
Molecular Weight (g/mol) 550.63
Formula Weight 550.63
Percent Purity 98.44%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.