missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ki67/MKI67 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-170DL594
This item is not returnable.
View return policy
Description
Ki67/MKI67 Polyclonal antibody specifically detects Ki67/MKI67 in Human, Mouse, Rat, Porcine, Avian samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay.
Specifications
| Ki67/MKI67 | |
| Polyclonal | |
| DyLight 594 | |
| PBS with 0.005% Sodium Azide | |
| antigen identified by monoclonal Ki-67, antigen KI-67, Antigen Ki67, antiKi67, Ki67, Ki-67, Ki67 ihc, Ki-67 ihc, Ki67 mouse, Ki-67 mouse, Ki67 western blot, Ki-67 western blot, KIA, Marker Of Proliferation Ki-67, MIB-, MIB-1, PPP1R105, Proliferation Marker Protein Ki-67, proliferation-related Ki-67 antigen, Protein Phosphatase 1, Regulatory Subunit 105 | |
| Rabbit | |
| Affinity Purified | |
| Breast Cancer, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Mitotic Regulators, Neuroscience | |
| 4288 | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay | |
| 1.0 mg/mL | |
| Western Blot 1-2ug/ml, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:50-1:300, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunohistochemistry-Paraffin 1:50-1:300, Immunohistochemistry-Frozen 1:50-1:300, Immunoblotting, Knockout Validated | |
| P46013, Q91VE6 | |
| MKI67 | |
| The immunogen for this KI67/MKI67 Antibody was made using a synthetic peptide from the internal region of Human KI67/MKI67, between amino acids: 1550-1600 [Uniprot: P46013]. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Pig, Avian | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction