missing translation for 'onlineSavingsMsg'
Learn More

Ki16425, MedChemExpress

Product Code. 16438317
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16438317

Brand: Medchem Express HY13285

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ki16425 (Debio 0719) is a subtype-selective, competitive antagonist of the EDG-family receptors, LPA1 and LPA3 with Kis of 0.34 μM and 0.93 μM, respectively. Ki16425 (Debio 0719) reduces the LPA-induced activation of p42/p44 MAPK. Ki16425 can also inhibit LPA-induced dephosphorylation of Yes-associated protein (YAP)/TAZ in HEK293A cells.

Ki16425 (Debio 0719) is a subtype-selective, competitive antagonist of the EDG-family receptors, LPA1 and LPA3 with Kis of 0.34 μM and 0.93 μM, respectively. Ki16425 (Debio 0719) reduces the LPA-induced activation of p42/p44 MAPK. Ki16425 can also inhibit LPA-induced dephosphorylation of Yes-associated protein (YAP)/TAZ in HEK293A cells.

Specifications

Chemical Name or Material Ki16425
Physical Form Solid
CAS 355025-24-0
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C23H23ClN2O5S
For Use With (Application) Cancer-Kinase/protease
Synonym Debio 0719
Solubility Information DMSO : ≥ 100 mg/mL (210.54 mM)
SMILES ClC1=C(C=CC=C1)C(C)OC(NC2=C(ON=C2C)C3=CC=C(C=C3)CSCCC(O)=O)=O
Molecular Weight (g/mol) 474.96
Formula Weight 474.96
Percent Purity 98.11%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.