missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ki-67 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28074-1-AP-150UL
This item is not returnable.
View return policy
Description
The Ki-67 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse Ki-67. This antibody recognizes mouse antigen. The Ki-67 antibody has been validated for the following applications: IHC, IF/ICC, IF-P, IF-Fro, FC (Intra), ELISA analysis.Specifications
| Ki-67 | |
| Polyclonal | |
| Unconjugated | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Mouse | |
| Polyclonal Antibody | |
| IgG |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA | |
| 400 μg/mL | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 17345 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction