missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KHSRP Recombinant Rabbit Monoclonal Antibody (23GB6435), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | KHSRP |
|---|---|
| Clone | 23GB6435 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088080
![]() |
Invitrogen™
MA553960 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088454
|
Invitrogen™
MA553961 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KHSRP Recombinant Monoclonal antibody specifically detects KHSRP in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| KHSRP | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q3U0V1, Q92945, Q99PF5 | |
| 16549, 171137, 8570 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6435 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 6330409F21Rik; Far upstream element-binding protein 2; FBP2; Fubp2; FUSE binding protein 2; FUSE-binding protein 2; KH type-splicing regulatory protein; KHE3; KHSRP; KH-type splicing regulatory protein; KH-type splicing regulatory protein (FUSE binding protein 2); KSRP; MAP2 RNA trans-acting protein 1; MAP2 RNA trans-acting protein MARTA1; MARTA1; p75 | |
| A synthesized peptide derived from human KHSRP (50-100AA). | |
| Primary | |
| -20°C | |
| KHSRP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title