missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KHSRP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55409-1-AP-150UL
This item is not returnable.
View return policy
Description
The KHSRP gene encodes a multifunctional RNA-binding protein implicated in a variety of cellular processes, including transcription, alternative pre-mRNA splicing, and mRNA localization.Specifications
| KHSRP | |
| Polyclonal | |
| Unconjugated | |
| KHSRP | |
| FBP2, FUBP2, FUSE binding protein 2, KHSRP, KSRP, p75 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16549, 171137, 8570 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.52 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3U0V1, Q92945, Q99PF5 | |
| Khsrp | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction