missing translation for 'onlineSavingsMsg'
Learn More

KHSRP Antibody (2G5), Novus Biologicals™

Product Code. 18335499 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18335499 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18335499 Supplier Novus Biologicals Supplier No. H00008570M05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KHSRP Monoclonal antibody specifically detects KHSRP in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen KHSRP
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2G5
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_003676
Gene Alias FBP2far upstream element-binding protein 2, FUBP2p75, FUSE binding protein 2, FUSE-binding protein 2, KH type-splicing regulatory protein, KH-type splicing regulatory protein, KSRPMGC99676
Host Species Mouse
Immunogen KHSRP (NP_003676, 151 a.a. ~ 239 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VPDGMVGLIIGRGGEQINKIQQDSGCKVQISPDSGGLPERSVSLTGAPESVQKAKMMLDDIVSRGRGGPPGQFHDNANGGQNGTVQEIM
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8570
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.