missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KHK Monoclonal Antibody (OTI1D8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525136
This item is not returnable.
View return policy
Description
KHK Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
KHK encodes the gene ketohexokinase that catalyzes conversion of fructose to fructose-1-phosphate. The splice variant presented encodes the highly active form found in liver. renal cortex. and small intestine. while the alternate variant encodes the lowe.
Specifications
| KHK | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P50053 | |
| KHK | |
| Full length human recombinant protein of KHK produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI1D8 | |
| Unconjugated | |
| KHK | |
| fructokinase; hepatic fructokinase; KETHPRO; ketohexokinase; ketohexokinase (fructokinase); ketohexokinase isoform a; Khk; testicular tissue protein Li 102 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3795 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction