missing translation for 'onlineSavingsMsg'
Learn More

Khellin, MP Biomedicals

Product Code. 11400936 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11400936 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11400936 Supplier MP Biomedicals Supplier No. 0520209210
 Restricted Item
Estimated Shipment: 29-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 82-02-0
Molecular Formula C14H12O5
Quantity 10 g
Synonym khellin, visammin, amicardine, methafrone, coronin, amikellin, ammipuran, ammivisnagen, benecardin, corafurone
InChI Key HSMPDPBYAYSOBC-UHFFFAOYSA-N
SMILES CC1=CC(=O)C2=C(C3=C(C(=C2O1)OC)OC=C3)OC
IUPAC Name 4,9-dimethoxy-7-methylfuro[3,2-g]chromen-5-one
Molecular Weight (g/mol) 260.245
PubChem CID 3828
ChEBI CHEBI:6133
Formula Weight 260.24
Chemical Name or Material Khellin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.