missing translation for 'onlineSavingsMsg'
Learn More

KH7, MedChemExpress

Product Code. 16496527
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16496527

Brand: Medchem Express HY103194

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress KH7 is a soluble adenylyl cyclase (sAC)-specific inhibitor, with IC50s of 3-10 μM toward both recombinant purified human sACt protein and heterologously expressed sACt in cellular assays. KH7 is also a cAMP inhibitor.

KH7 is a soluble adenylyl cyclase (sAC)-specific inhibitor, with IC50s of 3-10 μM toward both recombinant purified human sACt protein and heterologously expressed sACt in cellular assays. KH7 is also a cAMP inhibitor.

Specifications

Chemical Name or Material KH7
CAS 330676-02-3
Color White
Purity Grade Notes Research
Molecular Formula C17H15BrN4O2S
Quantity 5 mg
Solubility Information DMSO : 250 mg/mL (596.23 mM; Need ultrasonic)
SMILES CC(SC1=NC2=CC=CC=C2N1)C(N/N=C/C3=CC(Br)=CC=C3O)=O
Molecular Weight (g/mol) 419.3
Formula Weight 419.3
Percent Purity 98.06%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.