missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ KH 7
GREENER_CHOICE

Product Code. 18035912
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18035912

Brand: Tocris Bioscience™ 3834/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective soluble adenylyl cyclase inhibitor

Selective soluble adenylyl cyclase (sAC) inhibitor (IC50 = 3 - 10 μM in vivo). Inert towards transmembrane adenylyl cyclase (tmAC) in vitro and in whole cells at concentrations up to 300 μM. Blocks synthesis of cAMP and displays an antiapoptotic effect at concentrations of 1 - 100 μM.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 330676-02-3
Molecular Formula C17H15BrN4O2S

Specifications

Quantity 50 mg
Formula Weight 419.3
Purity >99%
Molecular Weight (g/mol) 419.297
Chemical Name or Material KH 7
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.