Learn More
PeproTech KGF Polyclonal Antibody, PeproTech®, Invitrogen™

Description
AA Sequence of recombinant protein: CNDMTPEQMA TNVNCSSPER HTRSYDYMEG GDIRVRRLFC RTQWYLRIDK RGKVKGTQEM KNNYNIMEIR TVAVGIVAIK GVESEFYLAM NKEGKLYAKK ECNEDCNFKE LILENHYNTY ASAKWTHNGG EMFVALNQKG IPVRGKKTKK EQKTAHFLPM AIT. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human KGF (FGF-7). Anti-Human KGF (FGF-7)-specific antibody was purified by affinity chromatography employing an immobilized Human KGF (FGF-7) matrix. Sandwich ELISA: To detect Human KGF (FGF-7) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human KGF (FGF-7) (500-P19Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human KGF (FGF-7). Western Blot: To detect Human KGF (FGF-7) by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human KGF (FGF-7) is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Specifications
Specifications
| Antigen | KGF |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | FGF7 |
| Gene Accession No. | P21781 |
| Gene Alias | Fgf7; FGF-7; Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; keratinocyte growth factor; Kgf |
| Gene Symbols | FGF7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.