Learn More
PeproTech KGF Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody
Brand: PeproTech 500-P19BT-50UG
Description
AA Sequence of recombinant protein: CNDMTPEQMA TNVNCSSPER HTRSYDYMEG GDIRVRRLFC RTQWYLRIDK RGKVKGTQEM KNNYNIMEIR TVAVGIVAIK GVESEFYLAM NKEGKLYAKK ECNEDCNFKE LILENHYNTY ASAKWTHNGG EMFVALNQKG IPVRGKKTKK EQKTAHFLPM AIT. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human KGF (FGF-7). Anti-Human KGF (FGF-7)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hKGF by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human KGF (500-P19) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hKGF. Western Blot: To detect Human KGF by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human KGF is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P19BT-1MG will be provided as 2 x 500 μg
Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation. Required for normal branching morphogenesis. Growth factor active on keratinocytes. Possible major paracrine effector of normal epithelial cell proliferation.
Specifications
| KGF | |
| Polyclonal | |
| Biotin | |
| FGF7 | |
| Fgf7; FGF-7; Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; keratinocyte growth factor; Kgf | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2252 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P21781 | |
| FGF7 | |
| E.coli-derived Recombinant Human KGF (FGF-7). | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.