missing translation for 'onlineSavingsMsg'
Learn More

KGF/FGF-7 Antibody (29522) [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 30737022 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737022 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737022 Supplier Novus Biologicals Supplier No. FAB251I

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KGF/FGF-7 Monoclonal antibody specifically detects KGF/FGF-7 in Human samples. It is validated for ELISA Standard (Matched Pair),Neutralization,ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair)
TRUSTED_SUSTAINABILITY

Specifications

Antigen KGF/FGF-7
Applications ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Classification Monoclonal
Clone 29522
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias FGF-7, fibroblast growth factor 7HBGF-7, Heparin-binding growth factor 7, keratinocyte growth factor, KGFfibroblast growth factor 7 (keratinocyte growth factor)
Host Species Mouse
Immunogen E. coli-derived recombinant human KGF/FGF-7, Cys32-Thr194, Accession # P21781
Purification Method Protein A or G purified from ascites
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2252
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.