missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KGF/FGF-7 Antibody (001), Alexa Fluor™ 647, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90560AF647
This item is not returnable.
View return policy
Description
KGF/FGF-7 Monoclonal specifically detects KGF/FGF-7 in Mouse samples. It is validated for ELISA.
Specifications
| KGF/FGF-7 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| FGF-7, fibroblast growth factor 7HBGF-7, Heparin-binding growth factor 7, keratinocyte growth factor, KGFfibroblast growth factor 7 (keratinocyte growth factor) | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse KGF/FGF-7 (NP_032034.1; Cys32-Thr194). | |
| 0.1 mL | |
| Angiogenesis, Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Markers | |
| 2252 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 1 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction