missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ KG 5

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
KG 5 is a PDGFRβ and B-Raf allosteric inhibitor. Also inhibits c-Raf, FLT3 and KIT. Disrupts blood vessel formation in zebrafish embryos and inhibits angiogenesis in a mouse angiogenesis model. Inhibits growth of renal and pancreatic tumors in mice. Also arrests cells in prometaphase. Orally available.
Specifications
Specifications
| Chemical Name or Material | 2-(Methylthio)-6-[4-[5-[[3-(trifluoromethyl)phenyl]amino]-1H-1,2,4-triazol-3-yl]phenoxy]-4-pyrimidinamine |
| CAS | 877874-85-6 |
| Quantity | 10 mg |
| Target | Raf Kinase Inhibitors |
| Molecular Formula | C20H16F3N7OS |
| Formula Weight | 459.45 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction