missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571424
This item is not returnable.
View return policy
Description
KF1 Polyclonal Antibody for Western Blot
The protein encoded by this gene contains a RING-H2 finger, a motif known to be involved in protein-protein and protein-DNA interactions. This gene is highly expressed in normal cerebellum, but not in the cerebral cortex. The expression of the rat counterpart in the frontal cortex and hippocampus was shown to be induced by elctroconvulsive treatment (ECT) as well as chronic antidepressant treatment, suggesting that this gene may be a molecular target for ECT and antidepressants. [provided by RefSeq].
Specifications
| KF1 | |
| Polyclonal | |
| Unconjugated | |
| RNF103 | |
| Adrg34; E3 ubiquitin-protein ligase RNF103; hKF-1; KF1; KF-1; RING finger protein 103; RING-type E3 ubiquitin transferase RNF103; RNF103; Zfp103; Zfp-103; zinc finger protein 103; zinc finger protein 103 homolog; zinc finger protein expressed in cerebellum | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 7844 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O00237 | |
| RNF103 | |
| synthetic peptide directed towards the middle region of human KF1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction