missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ KF 38789

Product Code. 18093802
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18093802 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18093802 Supplier R&D Systems™ Supplier No. 2748/10
  • ONLINE EXCLUSIVE PRICE
    £235.00
    £221.55 / 10mg
    Save £13.45
    5% Off
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective inhibitor of P-selectin-mediated cell adhesion

Selective inhibitor of P-selectin-mediated cell adhesion (IC50 = 1.97 μM) that displays no effects on L-selectin- and E-selectin-mediated adhesion. Blocks P-selectin-mediated binding in vitro and leukocyte accumulation in vivo.
TRUSTED_SUSTAINABILITY

Specifications

CAS 257292-29-8
Quantity 10 mg
Product Line Tocris
Molecular Formula C19H21NO5S
Formula Weight 375.44
Purity >97%
Molecular Weight (g/mol) 375.439
Chemical Name or Material KF 38789
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.