missing translation for 'onlineSavingsMsg'
Learn More

Ketotifen N-Oxide Hydrochloride, TRC

Product Code. 30396149
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30396149 500 mg
30405350 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30396149 Supplier Toronto Research Chemicals Supplier No. TRCK315410500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59743-91-8
Molecular Formula C19H19NO2S . HCl
Molecular Weight (g/mol) 361.88
Synonym 10H-Benzo[4,5]cyclohepta[1,2-b]thiophen-10-one, 4,9-dihydro-4-(1-methyl-1-oxido-4-piperidinylidene)-, hydrochloride (9CI), 10H-Benzo[4,5]cyclohepta[1,2-b]thiophen-10-one, 4,9-dihydro-4-(1-methyl-4-piperidinylidene)-, N-oxide, hydrochloride
IUPAC Name 1-methyl-4-(10-oxo-9,10-dihydro-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-4-ylidene)piperidine 1-oxide hydrochloride
SMILES O=C1C2=C(C=CS2)/C(C3=C(C=CC=C3)C1)=C4CCN(CC/4)(C)=O.Cl

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C19H19NO2S.ClH/c1-20(22)9-6-13(7-10-20)18-15-5-3-2-4-14(15)12-17(21)19-16(18)8-11-23-19;/h2-5,8,11H,6-7,9-10,12H2,1H3;1H/b18-13-;
Formula Weight 361.1
Chemical Name or Material Ketotifen N-Oxide Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.