missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ketorolac Trometamol, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Ketorolac trometamol |
| CAS | 74103-07-4 |
| Analyte or Component Names | Ketorolac trometamol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H13 N O3 . C4 H11 N O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ketorolac Trometamol, 1H-Pyrrolizine-1-carboxylic acid, 5-benzoyl-2,3-dihydro-, compd. with 2-amino-2-(hydroxymethyl)-1,3-propanediol (1:1), 1H-Pyrrolizine-1-carboxylic acid, 5-benzoyl-2,3-dihydro-, (+/-)-, compd. with 2-amino-2-(hydroxymethyl)-1,3-propanediol (1:1), 1,3-Propanediol, 2-amino-2-(hydroxymethyl)-, (+/-)-5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylate (1:1) (salt), 1,3-Propanediol, 2-amino-2-(hydroxymethyl)-, benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylate (1:1) (salt) (9CI), Acular, Acular PF, Acuvail, Dolac, ISV 304, Ketanov, Ketolac, Ketorol, Ketorolac tromethamine, Lixidol, Tarazyn, Toradol, Toratex, Trometamol Ketorolac, ketorolac tris salt |
| InChI Formula | InChI=1S/C15H13NO3.C4H11NO3/c17-14(10-4-2-1-3-5-10)13-7-6-12-11(15(18)19)8-9-16(12)13;5-4(1-6,2-7)3-8/h1-7,11H,8-9H2,(H,18,19);6-8H,1-3,5H2 |
| Solution Type | Neat |
| SMILES | NC(CO)(CO)CO.OC(=O)C1CCn2c1ccc2C(=O)c3ccccc3 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction