missing translation for 'onlineSavingsMsg'
Learn More

Ketorolac Ethyl Ester (Racemic), TRC

Product Code. 30386654
Change view
Click to view available options
Quantity:
25 mg
50 mg
200 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386654 200 mg
30377016 25 mg
30383082 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386654 Supplier Toronto Research Chemicals Supplier No. TRCE937730200MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108061-03-6
Molecular Formula C17 H17 N O3
Molecular Weight (g/mol) 283.32
Synonym 5-Benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylic acid ethyl ester, 1H-Pyrrolizine-1-carboxylic acid, 5-benzoyl-2,3-dihydro-, ethyl ester, (±)-, Ketorolac ethyl ester
IUPAC Name ethyl 5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylate
SMILES CCOC(=O)C1CCn2c1ccc2C(=O)c3ccccc3

Specifications

Quantity 200 mg
InChI Formula InChI=1S/C17H17NO3/c1-2-21-17(20)13-10-11-18-14(13)8-9-15(18)16(19)12-6-4-3-5-7-12/h3-9,13H,2,10-11H2,1H3
Formula Weight 283.1208
Chemical Name or Material Ketorolac Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.