missing translation for 'onlineSavingsMsg'
Learn More

Ketorolac-d5, TRC

Product Code. 30392122
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392122 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30392122 Supplier Toronto Research Chemicals Supplier No. TRCK23561710MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ketorolac-d5
CAS 1215767-66-0
Purity Grade Notes HPLC
Molecular Formula C15H8D5NO3
Quantity 10 mg
Synonym (±)-5-Benzoyl-d5-2,3-dihydro-1H-pyrrolizine-1-carboxylic Acid, (±)-Ketorolac-d5, Acular LS-d5, Keromin-d5, RS 37619-d5, Sprix-d5
InChI Formula InChI=1S/C15H13NO3/c17-14(10-4-2-1-3-5-10)13-7-6-12-11(15(18)19)8-9-16(12)13/h1-7,11H,8-9H2,(H,18,19)/i1D,2D,3D,4D,5D
SMILES O=C(C1=C([2H])C([2H])=C([2H])C([2H])=C1[2H])C2=CC=C3N2CCC3C(O)=O
Molecular Weight (g/mol) 260.3
Percent Purity >95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.