missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ketorolac 1,2-Propylene Glycol Esters (Mixture of Regio- and Stereoisomers), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Ketorolac 1,2-Propylene Glycol Esters (Mixture of Regio- and Stereoisomers)
Specifications
Specifications
| Chemical Name or Material | Ketorolac 1,2-Propylene Glycol Esters (Mixture of Regio- and Stereoisomers) |
| Analyte or Component Names | Ketorolac 1,2-Propylene Glycol Esters (Mixture of Regio- and Stereoisomers) |
| Shipping Condition | Room Temperature |
| Molecular Formula | 2 C18 H19 N O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ketorolac 1,2-Propylene Glycol Esters (Mixture of Regio- and Stereoisomers), Ketorolac 1,2-Propylene Glycol Ester (Mixture of Isomers) |
| InChI Formula | InChI=1S/2C18H19NO4/c1-12(20)11-23-18(22)14-9-10-19-15(14)7-8-16(19)17(21)13-5-3-2-4-6-13;1-12(11-20)23-18(22)14-9-10-19-15(14)7-8-16(19)17(21)13-5-3-2-4-6-13/h2*2-8,12,14,20H,9-11H2,1H3 |
| SMILES | CC(O)COC(=O)C1CCn2c1ccc2C(=O)c3ccccc3.CC(CO)OC(=O)C4CCn5c4ccc5C(=O)c6ccccc6 |
| IUPAC Name | 1-hydroxypropan-2-yl 5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylate;2-hydroxypropyl 5-benzoyl-2,3-dihydro-1H-pyrrolizine-1-carboxylate |
| Molecular Weight (g/mol) | 626.7 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction