missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ketoprofen EP Impurity D (Ketoprofen USP Related Compound A), Mikromol™
Description
Ketoprofen EP Impurity D (Ketoprofen USP Related Compound A)
Specifications
Specifications
| Chemical Name or Material | (2RS)-2-(3-(4-Methylbenzoyl)phenyl)propanoic Acid |
| CAS | 107257-20-5 |
| Analyte or Component Names | (2RS)-2-(3-(4-Methylbenzoyl)phenyl)propanoic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H16 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | (2RS)-2-[3-(4-Methylbenzoyl)phenyl]propanoic Acid, Ketoprofen Imp. D (EP), Ketoprofen Related Compound A, Ketoprofen Impurity D |
| InChI Formula | InChI=1S/C17H16O3/c1-11-6-8-13(9-7-11)16(18)15-5-3-4-14(10-15)12(2)17(19)20/h3-10,12H,1-2H3,(H,19,20) |
| SMILES | CC(C(=O)O)c1cccc(c1)C(=O)c2ccc(C)cc2 |
| IUPAC Name | 2-[3-(4-methylbenzoyl)phenyl]propanoic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?