missing translation for 'onlineSavingsMsg'
Learn More

(±)-Ketoprofen-d4 (propionic-d4 acid), TRC

Product Code. 30392861
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392861 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392861 Supplier Toronto Research Chemicals Supplier No. TRCK2008015MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (±)-Ketoprofen propionic-d4 acid
CAS 1219805-29-4
Molecular Formula C16 2H4 H10 O3
Quantity 5 mg
Synonym (±)-Ketoprofen D4 (propionic D4 acid), 2-(3-Benzoylphenyl)-2,3,3,3-tetradeuterio-propanoic acid
InChI Formula InChI=1S/C16H14O3/c1-11(16(18)19)13-8-5-9-14(10-13)15(17)12-6-3-2-4-7-12/h2-11H,1H3,(H,18,19)/i1D3,11D
SMILES [2H]C([2H])([2H])C([2H])(C(=O)O)c1cccc(c1)C(=O)c2ccccc2
IUPAC Name 2-(3-benzoylphenyl)-2,3,3,3-tetradeuteriopropanoic acid
Molecular Weight (g/mol) 258.31
Formula Weight 258.1194
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.