missing translation for 'onlineSavingsMsg'
Learn More

Ketoconazole, 99.17%, MP Biomedicals™

Product Code. 11491371 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491371 50 mg 50mg
11401381 100 mg 100mg
11411381 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491371 Supplier MP Biomedicals Supplier No. 0215915850
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 65277-42-1 | Formula: C26H28Cl2N4O4 | Weight: 534.452 g/mol

Chemical Identifiers

CAS 65277-42-1
Molecular Formula C26H28Cl2N4O4
Molecular Weight (g/mol) 534.452
MDL Number MFCD00058579
InChI Key XMAYWYJOQHXEEK-SIULDFEJSA-N
Synonym cis-1-Acetyl-4-(4-((2-(2, 4-dichlorophenyl)-2-(1H-1M-idazol-1-ylmethyl)-1, 3-dioxolan-4-yl) methoxy) phenyl)-piperazine, Ketoisdin
PubChem CID 76973198
IUPAC Name 2,2,2-trideuterio-1-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]ethanone
SMILES CC(=O)N1CCN(CC1)C2=CC=C(C=C2)OCC3COC(O3)(CN4C=CN=C4)C5=C(C=C(C=C5)Cl)Cl

Specifications

Type Ketoconazole
Physical Form Powder
Color White
Quantity 50 mg
Assay Percent Range >99%
Formula Weight 531.44
Chemical Name or Material Ketoconazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.