missing translation for 'onlineSavingsMsg'
Learn More

Ketoconazole, 98%

Product Code. 15425619
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15425619 25 g 25g
15415619 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15425619 Supplier Thermo Scientific Alfa Aesar Supplier No. J63367.14
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent inhibitor of cytochrome P450c17 | CAS: 65277-42-1 | C26H28Cl2N4O4 | 534.452 g/mol

Ketoconazole is used to treat candidiasis, chronic mucocutaneous candidiasis, oral thrush, candiduria, blastomycosis, coccidioidomycosis, histoplasmosis, chromomycosis, and paracoccidioidomycosis. Ketoconazole is an antifungal agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ketoconazole is used to treat candidiasis, chronic mucocutaneous candidiasis, oral thrush, candiduria, blastomycosis, coccidioidomycosis, histoplasmosis, chromomycosis, and paracoccidioidomycosis. Ketoconazole is an antifungal agent.

Solubility
Soluble in DMSO, ethanol, chloroform, water, and methanol.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65277-42-1
Molecular Formula C26H28Cl2N4O4
Molecular Weight (g/mol) 534.452
MDL Number MFCD00058579
InChI Key XMAYWYJOQHXEEK-SIULDFEJSA-N
Synonym ketoconazole
PubChem CID 76973198
IUPAC Name 2,2,2-trideuterio-1-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]ethanone
SMILES CC(=O)N1CCN(CC1)C2=CC=C(C=C2)OCC3COC(O3)(CN4C=CN=C4)C5=C(C=C(C=C5)Cl)Cl

Specifications

Color White
Quantity 25 g
UN Number UN2811
Merck Index 14,5302
Solubility Information Soluble in DMSO,ethanol,chloroform,water,and methanol.
Formula Weight 531.43
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Ketoconazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.