missing translation for 'onlineSavingsMsg'
Learn More

Keto Pioglitazone (M-III), TRC

Product Code. 30400254
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400254 10 mg
30397986 1 mg
30389387 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400254 Supplier Toronto Research Chemicals Supplier No. TRCK20060010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146062-45-5
Molecular Formula C19 H18 N2 O4 S
Molecular Weight (g/mol) 370.42
Synonym 5-[[4-[2-(5-acetyl-2-pyridyl)ethoxy]phenyl]methyl]thiazolidine-2,4-dione, 2,4-Thiazolidinedione, 5-[[4-[2-(5-acetyl-2-pyridinyl)ethoxy]phenyl]methyl]-
IUPAC Name 5-[[4-[2-(5-acetylpyridin-2-yl)ethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione
SMILES CC(=O)c1ccc(CCOc2ccc(CC3SC(=O)NC3=O)cc2)nc1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C19H18N2O4S/c1-12(22)14-4-5-15(20-11-14)8-9-25-16-6-2-13(3-7-16)10-17-18(23)21-19(24)26-17/h2-7,11,17H,8-10H2,1H3,(H,21,23,24)
Formula Weight 370.0987
Chemical Name or Material keto Pioglitazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.