missing translation for 'onlineSavingsMsg'
Learn More

Keto Itraconazole, TRC

Product Code. 30405040
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30405040 5 mg -
30389394 10 mg -
30393508 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30405040 Supplier Toronto Research Chemicals Supplier No. TRCK1950005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112560-33-5
Molecular Formula C35 H36 Cl2 N8 O5
Molecular Weight (g/mol) 719.62
Synonym 3H-1,2,4-Triazol-3-one, 4-[4-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]-1-piperazinyl]phenyl]-2,4-dihydro-2-(1-methyl-2-oxopropyl)-, rel-, 3H-1,2,4-Triazol-3-one, 4-[4-[4-[4-[[2-(2,4-dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]-1-piperazinyl]phenyl]-2,4-dihydro-2-(1-methyl-2-oxopropyl)-, (2α,4α)-(±)-, rel-4-[4-[4-[4-[[(2R,4S)-2-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]-1-piperazinyl]phenyl]-2,4-dihydro-2-(1-methyl-2-oxopropyl)-3H-1,2,4-triazol-3-one, 3H-1,2,4-Triazol-3-one, 4-[4-[4-[4-[[2-(2,4-dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]-1-piperazinyl]phenyl]-2,4-dihydro-2-(1-methyl-2-oxopropyl)-, (2α,4α)-, Keto itraconazole
IUPAC Name 4-[4-[4-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazin-1-yl]phenyl]-2-(3-oxobutan-2-yl)-1,2,4-triazol-3-one
SMILES CC(N1N=CN(C1=O)c2ccc(cc2)N3CCN(CC3)c4ccc(OC[C@H]5CO[C@@](Cn6cncn6)(O5)c7ccc(Cl)cc7Cl)cc4)C(=O)C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C35H36Cl2N8O5/c1-24(25(2)46)45-34(47)44(23-40-45)29-6-4-27(5-7-29)41-13-15-42(16-14-41)28-8-10-30(11-9-28)48-18-31-19-49-35(50-31,20-43-22-38-21-39-43)32-12-3-26(36)17-33(32)37/h3-12,17,21-24,31H,13-16,18-20H2,1-2H3/t24?,31-,35-/m0/s1
Formula Weight 718.2186
Chemical Name or Material Keto Itraconazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.