missing translation for 'onlineSavingsMsg'
Learn More

Ketanserin Tartrate, TRC

Product Code. 30134500
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134500 1 g 1g
30136115 500 mg 500mg
30041323 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134500 Supplier Toronto Research Chemicals Supplier No. TRCK1654531G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83846-83-7
Molecular Formula C22 H22 F N3 O3 . C4 H6 O6
Molecular Weight (g/mol) 545.51
Synonym Ketanserin Hydrogen Tartrate, 3-[2-[4-(4-Fluorobenzoyl)-1-piperidinyl]ethyl]-2,4(1H,3H)-quinazolinedione (2R,3R)-2,3-dihydroxybutanedioate (1:1), 2,4(1H,3H)-Quinazolinedione, 3-[2-[4-(4-fluorobenzoyl)-1-piperidinyl]ethyl]-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1), KJK 945, Ket, Ketanserin tartrate, Perketan, R 49945, Serepress, Sufrexal
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;3-[2-[4-(4-fluorobenzoyl)piperidin-1-yl]ethyl]-1H-quinazoline-2,4-dione
SMILES O[C@H]([C@@H](O)C(=O)O)C(=O)O.Fc1ccc(cc1)C(=O)C2CCN(CCN3C(=O)Nc4ccccc4C3=O)CC2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C22H22FN3O3.C4H6O6/c23-17-7-5-15(6-8-17)20(27)16-9-11-25(12-10-16)13-14-26-21(28)18-3-1-2-4-19(18)24-22(26)29;5-1(3(7)8)2(6)4(9)10/h1-8,16H,9-14H2,(H,24,29);1-2,5-6H,(H,7,8)(H,9,10)/t;1-,2-/m.1/s1
Formula Weight 545.181
Chemical Name or Material Ketanserin Tartrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.