missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ketanserin tartrate

Product Code. 30001144
Change view
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001144 50 mg 50mg
18752664 250 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001144 Supplier Tocris Bioscience™ Supplier No. 0908/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective 5-HT2A antagonist. Also antagonist at 5-HT1D

Selective 5-HT2A serotonin receptor antagonist; can also be used to discriminate between 5-HT1D and 5-HT1B receptor subtypes.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83846-83-7
Molecular Formula C26H30FN3O10

Specifications

Quantity 50 mg
Formula Weight 545.52
Purity >97%
Molecular Weight (g/mol) 563.54
Chemical Name or Material Ketanserin tartrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.