missing translation for 'onlineSavingsMsg'
Learn More

Ketanserin tartrate, MedChemExpress

Product Code. 16495026
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495026 100 mg 100mg
16410407 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16495026 Supplier Medchem Express Supplier No. HY10562A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ketanserin (R41468) tartrate is a selective 5-HT2 receptor antagonist. Ketanserin tartrate also blocks hERG current (IhERG) in a concentration-dependent manner (IC50=0.11 μM).

Ketanserin (R41468) tartrate is a selective 5-HT2 receptor antagonist. Ketanserin tartrate also blocks hERG current (IhERG) in a concentration-dependent manner (IC50=0.11 μM).

Specifications

Chemical Name or Material Ketanserin tartrate
CAS 83846-83-7
Color White
Purity Grade Notes Research
Molecular Formula C26H28FN3O9
Quantity 100 mg
Synonym R41468 tartrate
Solubility Information DMSO : 150 mg/mL (274.97 mM; Need ultrasonic) ∣H2O : 6 mg/mL (11.00 mM; Need ultrasonic)
SMILES O=C(C1=CC=CC=C1N2)N(CCN3CCC(C(C4=CC=C(F)C=C4)=O)CC3)C2=O.O=C(O)[C@H](O)[C@@H](O)C(O)=O
Molecular Weight (g/mol) 545.51
Formula Weight 545.51
Percent Purity 99.49%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.