missing translation for 'onlineSavingsMsg'
Learn More

Kerosene, Reagent grade, Honeywell™ Riedel-de Haën™

Product Code. 15693860 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
3.8 L
18.9 L
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
18.9 L
3.8 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15693860 3.8 L Glass Bottle 3.8 L
15683860 18.9 L Stainless Steel Can 18.9 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15693860 Supplier Honeywell Research Chemicals Supplier No. 3294603.8L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

reagent grade, low odor

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8008-20-6
MDL Number MFCD00135561

Specifications

Color Colorless
Density 0.8g/mL at 25°C
Boiling Point 175°C to 325°C
Flash Point 70°C (158°F)
Packaging Glass Bottle
Quantity 3.8 L
UN Number UN1223
Vapor Density 4.5 (vs air)
Grade Reagent
Physical Form Liquid
Chemical Name or Material Kerosene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.