missing translation for 'onlineSavingsMsg'
Learn More

Kerosene, low odor

Product Code. p-7046470
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2.5 L
500mL
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 11323546

Brand: Thermo Scientific Alfa Aesar L14479.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 64742-47-8

Kerosene is ideal for dilution of oils and other organic fluids for determination of metal traces. It is used in paints & coatings, consumer products, printing inks and agricultural chemical applications. It is also used in food contact applications such as aluminum rolling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Kerosene is ideal for dilution of oils and other organic fluids for determination of metal traces. It is used in paints & coatings, consumer products, printing inks and agricultural chemical applications. It is also used in food contact applications such as aluminum rolling.

Solubility
Soluble in water (0.02 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64742-47-8
MDL Number MFCD00135561
Synonym Kerosine

Specifications

Density 0.8
Boiling Point 200°C to 250°C
Flash Point 76°C (169°F)
Refractive Index 1.444
Quantity 500 mL
Merck Index 14,5294
Solubility Information Soluble in water (0.02g/L at 20°C).
Chemical Name or Material Kerosene, low odor
compliance-icons
Product Identifier
  • Kerosene, low odor
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contains : Petroleum distillates, hydrotreated light

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.