Learn More
Invitrogen™ Keratocan Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143975
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: rat eye tissue, mouse eye tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Keratocan (KTN), also known as keratan sulfate proteoglycan keratocan, is a protein that in humans is encoded by the KERA gene. It is mapped to 12q22. The protein encoded by this gene is a keratan sulfate proteoglycan that is involved in corneal transparency. Defects in this gene are a cause of autosomal recessive cornea plana 2 (CNA2). Keratan sulfate proteoglycans (KSPGs) are members of the small leucine-rich proteoglycan (SLRP) family. KSPGs, particularly keratocan, lumican and mimecan, are important to the transparency of the cornea.
Specifications
| Keratocan | |
| Polyclonal | |
| Unconjugated | |
| Kera | |
| CNA2; KERA; Keratan sulfate proteoglycan keratocan; keratocan; Ktcn; KTN; SLRR2B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16545, 314771 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| O35367 | |
| Kera | |
| E.coli-derived mouse Keratocan/KTN/Kera recombinant protein (Position: R73-K115). | |
| 100 μg | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.