missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Keratinocyte-SFM (1X), with L-glutamine, without calcium chloride
GREENER_CHOICE

Product Code. 11504546
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 11504546

Brand: Gibco™ 10725018

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Keratinocyte-SFM (1X), with L-glutamine, without calcium chloride

With: L-Glutamine

Without: Calcium Chloride.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Keratinocyte Cells, Epithelial Cells
Classification Serum-free
Concentration 1 X
Form Liquid
Product Type Serum Free Medium (SFM)
Serum Level Serum-free
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives Glutamine, Phenol Red
Without Additives No Calcium Chloride
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Store in refrigerator (2–8°C).
Culture Type Mammalian Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.