missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Keratin Associated Protein 20-3 Polyclonal specifically detects Keratin Associated Protein 20-3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | Keratin Associated Protein 20-3 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | Q3LI60 |
| Gene Alias | KAP19D, KAP20.3, Keratin Associated Protein 19 Pseudogene 4, Keratin-Associated Protein 20-3, KRTAP19P4, KRTAP20-3 |
| Gene Symbols | KRTAP20-3 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?