missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Keratin 31 Antibody (LHTric-1), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53114AF350
This item is not returnable.
View return policy
Description
Keratin 31 Monoclonal antibody specifically detects Keratin 31 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Keratin 31 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Hair Keratin, Type I Ha, Hard Keratin, Type I, 1, hHa1, K31, Keratin 31, Keratin, Hair, Acidic, 1, Keratin, Type I Cuticular Ha1, Keratin-31, KRTHA1 | |
| CVPRPRCGPCNSFVR | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 3881 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| LHTric-1 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction