missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, Optima™ LC/MS Grade, Fisher Chemical™

Product Code. 10001334 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Unit Size:
1 L
2.5 L
4 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10001334 2.5 L 2.5 L
10055454 500 mL 500mL
10489553 1 L 1 L
10061044 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10001334 Supplier Fisher Chemical Supplier No. A955212
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Acetonitrile, >-99.9%, C2H3N, CAS Number-75-05-8, methane, cyano, ethyl nitrile, methanecarbonitrile, methylkyanid, mecn, acetonitrile, methyl cyanide, ethanenitrile, cyanure de methyl, cyanomethane, 1L | CAS: 75-05-8 | C2H3N | 41.053 g/mol

Acetonitrile, >-99.9%, C2H3N, CAS Number-75-05-8, methane, cyano, ethyl nitrile, methanecarbonitrile, methylkyanid, mecn, acetonitrile, methyl cyanide, ethanenitrile, cyanure de methyl, cyanomethane

  • Ideal mobile phase for LC/MS and UHPLC-UV applications in pharmaceutical, biotechnology, clinical, environmental and food safety industries.
  • Novel gradient elution test with Photodiode Array Detector screens for UV-absorbing contaminants at every wavelength in the 200 to 400nm range to afford smooth baselines and to reduce interferences.
  • Evaluated for 17 metal impurities at ppb concentrations to minimize the formation of metal adducts during analysis and simplify spectral interpretation.
  • 0.1 micron filtration minimizes troublesome particles that may interfere with LC/MS and UHPLC system performance
  • Provides ideal solution to prepare cleaner, particle-free mobile phase blends. Positive mode ionization (as propazine) and negative mode ionization (as mecoprop) with ESI and Single Quadrupole MS detect organic contaminants at max 50ppb and max 300ppb, respectively
  • Glass bottles have fluorinated lining
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Weight (g/mol) 41.053
MDL Number 1878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46°C
Absorbance ≤1.00 AU at 190nm, ≤0.15 AU at 195nm, ≤0.05 AU at 200nm, ≤0.04 AU at 205nm, ≤0.03 AU at 210nm, ≤0.025 AU at 215nm, ≤0.015 AU at 220nm, ≤0.015 AU at 225nm, ≤0.01 AU at 230nm, ≤0.005 AU at 254nm, ≤0.005 AU at 280nm
Color Colorless
Decomposition Information >500°C
Density 0.781g/cm³
Quantity 2.5 L
Identification Pass Test
Packaging Glass Bottle with fluorinated lining
Physical Form Liquid
Residue after Evaporation ≤0.8ppm
Assay Percent Range Assay: 99.9% min by GC
Titratable Acid ≤0.008meq/g
Molecular Formula C2H3N
Titratable Base ≤0.0006meq/g
Linear Formula CH3CN
Vapor Pressure 97mbar at 20°C
Viscosity 0.36cp at 20°C
UN Number 1648
Water ≤0.01%
Merck Index 15, 69
Boiling Point 81°C
Formula Weight 41.05
Percent Purity ≥99.9%
Grade Formulated for UHPLC-UV
Assay GC
Chemical Name or Material Acetonitrile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Acute toxicity Category 4
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Acetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.