Learn More
KELL Monoclonal Antibody (BRIC203), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA180068
Code : LD
Additional Details : Weight : 0.01000kg
Description
In haemaggulation tests, MA1-80068 reacts with Kp (a-b+) but not Kp (a+b-). This antibody will agglutinate Kp (a+b-c+) and Kp (a-b-c+) erythrocytes but will not react with Ko erythrocytes. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD238 antibody, clone BRIC203 recognizes human CD238, also known as the kell blood group antigen Kpbc. The kell blood group antigens are carried on the approximately 93 kDa erythrocyte membrane glycoprotein.
This gene encodes a type II transmembrane glycoprotein that is the highly polymorphic Kell blood group antigen. The Kell glycoprotein links via a single disulfide bond to the XK membrane protein that carries the Kx antigen. The encoded protein contains sequence and structural similarity to members of the neprilysin (M13) family of zinc endopeptidases.Specifications
KELL | |
Monoclonal | |
1 mg/mL | |
TBS with ≤100mM glycine and <0.1% sodium azide | |
P23276 | |
KEL | |
Human erythrocytes. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Hemagglutination Assay, Immunoprecipitation | |
BRIC203 | |
Unconjugated | |
KEL | |
CD238; ECE3; Kel; Kel blood group protein; Kell blood group; kell blood group antigen; kell blood group glycoprotein; Kell blood group glycoprotein homolog; Kell blood group, metallo-endopeptidase; Kell protein; RGD1565470 | |
Mouse | |
Protein G | |
RUO | |
3792 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |