missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Kell Antibody (4B10), Novus Biologicals™

Product Code. 18346858 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18346858 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18346858 Supplier Novus Biologicals™ Supplier No. H00003792M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Kell Monoclonal antibody specifically detects Kell in Human samples. It is validated for ELISA,Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kell
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4B10
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_000411
Gene Alias CD238, CD238 antigen, EC 3.4.24.-, ECE3, Kell blood group, kell blood group antigen, kell blood group glycoprotein, Kell blood group, metalloendopeptidase, Kell blood group, metallo-endopeptidase
Host Species Mouse
Immunogen KEL (NP_000411.1, 633 a.a. ∽ 732 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LENAADVGGLAIALQAYSKRLLRHHGETVLPSLDLSPQQIFFRSYAQVMCRKPSPQDSHDTHSPPHLRVHGPLSSTPAFARYFRCARGALLNPSSRCQLW
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 3792
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.