missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kelch-Like Family Member 21 Antibody (2H4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37659
This item is not returnable.
View return policy
Description
Kelch-Like Family Member 21 Monoclonal specifically detects Kelch-Like Family Member 21 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Kelch-Like Family Member 21 | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| KLHL21 | |
| Purified recombinant fragment of human KLHL21 expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| 9903 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 2H4 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| kelch-like 21 (Drosophila), Kelch-Like Protein 21, KIAA0469, KLHL21 | |
| Mouse | |
| 66 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction